Gene Transcripts

Gene Symbol
TARBP2
Entrez Gene ID
6895
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Gene Aliases
LOQS, TRBP, TRBP1, TRBP2
Chromosome Location
Chr. 12: 53500921 - 53506431 on Build GRCh38
UniGene ID
Hs.326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004178.4 NP_004169.3 4 431
NM_134323.1 NP_599150.1 4 896
NM_134324.2 NP_599151.2 4 471
XM_005269114.1 XP_005269171.1 4 466
XM_005269115.2 XP_005269172.1 4 473
XM_005269117.1 XP_005269174.1 4 515
XM_005269120.4 XP_005269177.1 4 879
XM_005269122.3 XP_005269179.1 4 451
XM_006719581.1 XP_006719644.1 3 315
XM_011538712.2 XP_011537014.1 4 520
XM_017019910.1 XP_016875399.1 4 466
XM_017019911.1 XP_016875400.1 4 609
GenBank mRNA AK290737.1 - 4 469
AK309569.1 - 4 515
BC002419.2 - 4 879
BC005860.2 - 4 761 2960701
BI771121.1 - 4 474
BT007140.1 - 4 408
M60801.1 - 4 399
U08998.2 - 4 496

View Full Product Details