Gene Transcripts

Gene Symbol
Kif6
Entrez Gene ID
171291
Gene Name
kinesin family member 6
Gene Aliases
Krp3
Chromosome Location
Chr. 9: 13016973 - 13311951 on Build Rnor_6.0
UniGene ID
Rn.207571
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006244480.1 XP_006244542.1 7 782
GenBank mRNA AF035952.2 - 7 775
AY035403.1 - 7 654

View Full Product Details