Gene Transcripts

Gene Symbol
Cklf
Entrez Gene ID
245978
Gene Name
chemokine-like factor
Gene Aliases
Cklf1
Chromosome Location
Chr. 19: 904758 - 914880 on Build Rnor_6.0
UniGene ID
Rn.81197
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139111.1 NP_620811.1 3 437
GenBank mRNA AF253064.2 - 2 278
AF253065.2 - 3 437

View Full Product Details