Gene Transcripts

Gene Symbol
Gbx1
Entrez Gene ID
246149
Gene Name
gastrulation brain homeobox 1
Gene Aliases
Chromosome Location
Chr. 4: 7173961 - 7200752 on Build Rnor_6.0
UniGene ID
Rn.92432
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271453.1 NP_001258382.1 2 1414
GenBank mRNA AF219999.1 - 1 36

View Full Product Details