Gene Transcripts

Gene Symbol
Ly49i2
Entrez Gene ID
260324
Gene Name
Ly49 inhibitory receptor 2
Gene Aliases
Chromosome Location
Chr. 4: 164876252 - 164900873 on Build Rnor_6.0
UniGene ID
Rn.86417
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152848.1 NP_690061.1 4 440
GenBank mRNA AY115572.1 - 4 440

View Full Product Details