Gene Transcripts

Gene Symbol
Testin
Entrez Gene ID
286916
Gene Name
testin gene
Gene Aliases
Chromosome Location
Chr. 17: 4194795 - 4225376 on Build Rnor_6.0
UniGene ID
Rn.10029
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173132.2 NP_775155.1 2 187
XM_008771428.1 XP_008769650.1 3 547
GenBank mRNA U16858.1 - 2 191

View Full Product Details