Gene Transcripts
- Gene Symbol
- Ceacam10
- Entrez Gene ID
- 287009
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 10
- Gene Aliases
- Chromosome Location
- Chr. 1: 81643809 - 81650085 on Build Rnor_6.0
- UniGene ID
- Rn.92160
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173339.1 |
NP_775461.1
|
2 |
431 |
|
GenBank mRNA |
U23056.1 |
- |
2 |
431 |
|
- Gene Symbol
- LOC103689942
- Entrez Gene ID
- 103689942
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 1-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 83003835 - 83010148 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details