Gene Transcripts

Gene Symbol
TSPAN6
Entrez Gene ID
7105
Gene Name
tetraspanin 6
Gene Aliases
T245, TM4SF6, TSPAN-6
Chromosome Location
Chr. X: 100627108 - 100637104 on Build GRCh38
UniGene ID
Hs.43233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278740.1 NP_001265669.1 1 83
NM_003270.3 NP_003261.1 1 208
GenBank mRNA AB097016.1 - 1 104
AB209876.1 - 1 167
AF043906.1 - 1 112
AF053453.1 - 1 64
AF133426.1 - 1 71
AK299176.1 - 1 59
AK300852.1 - 1 202
AK309581.1 - 1 66
AK315773.1 - 1 189
AY358825.1 - 1 85
BC012389.1 - 1 148 3857537
BT006977.1 - 1 45
CR457115.1 - 1 45
CX760536.1 - 1 208
DA147330.1 - 1 83
U84895.1 - 1 112

View Full Product Details