Gene Transcripts

Gene Symbol
Wfikkn2
Entrez Gene ID
287631
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2
Gene Aliases
RGD1305361
Chromosome Location
Chr. 10: 81937239 - 81943144 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247173.2 XP_006247235.1 2 1138
XM_220855.8 XP_220855.1 2 1987

View Full Product Details