Gene Transcripts

Gene Symbol
Krtap15-1
Entrez Gene ID
288297
Gene Name
keratin associated protein 15-1
Gene Aliases
Krtap15
Chromosome Location
Chr. 11: 28780446 - 28781270 on Build Rnor_6.0
UniGene ID
Rn.218461
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105896.1 NP_001099366.1 1 324

View Full Product Details