Gene Transcripts

Gene Symbol
Zfp12
Entrez Gene ID
288486
Gene Name
zinc finger protein 12
Gene Aliases
RGD1564396
Chromosome Location
Chr. 12: 13370504 - 13384038 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751129.3 XP_003751177.1 4 747
XM_221917.8 XP_221917.5 5 843

View Full Product Details