Gene Transcripts

Gene Symbol
CRISP2
Entrez Gene ID
7180
Gene Name
cysteine rich secretory protein 2
Gene Aliases
CRISP-2, CT36, GAPDL5, TPX1, TSP1
Chromosome Location
Chr. 6: 49676665 - 49714198 on Build GRCh38
UniGene ID
Hs.2042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142407.2 NP_001135879.1 8 790
NM_001142408.2 NP_001135880.1 9 847
NM_001142417.2 NP_001135889.1 9 827
NM_001142435.2 NP_001135907.1 8 810
NM_001261822.1 NP_001248751.1 8 816
NM_003296.3 NP_003287.1 9 853
XM_005249349.1 XP_005249406.1 10 932
XM_005249350.1 XP_005249407.1 9 890
XM_005249351.3 XP_005249408.1 9 910
XM_005249352.3 XP_005249409.1 9 901
XM_005249353.3 XP_005249410.1 10 937
XM_005249356.1 XP_005249413.1 10 935
XM_011514841.1 XP_011513143.1 10 1267
XM_011514842.1 XP_011513144.1 9 1230
XM_011514843.2 XP_011513145.1 10 937
GenBank mRNA AK223337.1 - 9 790
AK292385.1 - 9 831
AY289796.1 - 9 797
AY292862.1 - 9 892
AY292863.1 - 8 791
AY773479.1 - 9 833
BC022011.2 - 9 800 4826427
BC107707.1 - 8 747 6618679
BI831679.1 - 3 202
BM552843.1 - 8 725
BP369727.1 - 3 304
M25532.1 - 9 776
X95239.1 - 9 827

View Full Product Details