Gene Transcripts

Gene Symbol
Kiss1
Entrez Gene ID
289023
Gene Name
KiSS-1 metastasis-suppressor
Gene Aliases
Eseptin
Chromosome Location
Chr. 13: 50529506 - 50535431 on Build Rnor_6.0
UniGene ID
Rn.66008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181692.1 NP_859043.1 1 92
XM_008769443.1 XP_008767665.1 2 545
GenBank mRNA AY196983.1 - 1 92
JX139030.1 - 2 503
JX139031.1 - 2 299
JX139032.1 - 2 224

View Full Product Details