Gene Transcripts

Gene Symbol
Dhx15
Entrez Gene ID
289693
Gene Name
DEAH (Asp-Glu-Ala-His) box helicase 15
Gene Aliases
Chromosome Location
Chr. 14: 61136742 - 61174880 on Build Rnor_6.0
UniGene ID
Rn.55597
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191597.1 NP_001178526.1 9 1702
XM_006251031.2 XP_006251093.1 9 1706

View Full Product Details