Gene Transcripts

Gene Symbol
Mrpl32
Entrez Gene ID
291206
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
Chromosome Location
Chr. 17: 53102534 - 53105400 on Build Rnor_6.0
UniGene ID
Rn.13489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106116.1 NP_001099586.1 1 124

View Full Product Details