Gene Transcripts

Gene Symbol
UBE2D1
Entrez Gene ID
7321
Gene Name
ubiquitin conjugating enzyme E2 D1
Gene Aliases
E2(17)KB1, SFT, UBC4/5, UBCH5, UBCH5A
Chromosome Location
Chr. 10: 58334979 - 58370753 on Build GRCh38
UniGene ID
Hs.129683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204880.1 NP_001191809.1 5 563
NM_003338.4 NP_003329.1 6 595
XM_017016609.1 XP_016872098.1 6 494
GenBank mRNA AF257220.1 - 6 518
AJ272367.1 - 6 518
AK291901.1 - 6 566
BC005980.1 - 6 566 4106710
BC015997.1 - 6 570 4305900
BI598342.1 - 6 600
BT007041.1 - 6 372
DA618122.1 - 5 536
X78140.1 - 6 387

View Full Product Details