Gene Transcripts
- Gene Symbol
- Ssty1
- Entrez Gene ID
- 301816
- Gene Name
- spermiogenesis specific transcript on the Y 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 182842652 - 182843326 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_229292.8 |
XP_229292.6
|
1 |
544 |
|
- Gene Symbol
- LOC102553316
- Entrez Gene ID
- 102553316
- Gene Name
- Y-linked testis-specific protein 1-like
- Gene Aliases
- Chromosome Location
- Chr. 3: 133479944 - 133480618 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details