Gene Transcripts

Gene Symbol
Vom2r26
Entrez Gene ID
292531
Gene Name
vomeronasal 2 receptor, 26
Gene Aliases
Gm1961
Chromosome Location
Chr. 1: 63535051 - 63571794 on Build Rnor_6.0
UniGene ID
Rn.217922
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099471.1 NP_001092941.1 3 662

View Full Product Details