Gene Transcripts

Gene Symbol
LOC100911519
Entrez Gene ID
100911519
Gene Name
p53-induced protein with a death domain-like
Gene Aliases
Chromosome Location
Chr. 1: 213981826 - 213988126 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749011.3 XP_003749059.1 12 3257
XM_006230592.2 XP_006230654.1 12 2807
XM_006230593.2 XP_006230655.1 11 3281
XM_008760030.1 XP_008758252.1 11 3329
XM_008760031.1 XP_008758253.1 12 2713

Gene Symbol
Pidd1
Entrez Gene ID
293625
Gene Name
p53-induced death domain protein 1
Gene Aliases
Lrdd, Pidd
Chromosome Location
Chr. 1: 214418732 - 214425032 on Build Rnor_6.0
UniGene ID
Rn.127674
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106318.2 NP_001099788.2 12 2184
XM_006230543.2 XP_006230605.1 12 2807
XM_006230544.2 XP_006230606.1 11 3281
XM_008759983.1 XP_008758205.1 11 3329
XM_008759984.1 XP_008758206.1 12 2713

View Full Product Details