Gene Transcripts

Gene Symbol
Mphosph10
Entrez Gene ID
293828
Gene Name
M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein)
Gene Aliases
Chromosome Location
Chr. 1: 125212869 - 125229060 on Build Rnor_6.0
UniGene ID
Rn.103228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106340.1 NP_001099810.1 2 305
GenBank mRNA BC161909.1 - 2 278

View Full Product Details