Gene Transcripts

Gene Symbol
Tpd52l3
Entrez Gene ID
293894
Gene Name
tumor protein D52-like 3
Gene Aliases
RGD1309391, Trpd52l3
Chromosome Location
Chr. 1: 248194680 - 248198193 on Build Rnor_6.0
UniGene ID
Rn.225788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106349.1 NP_001099819.1 1 194
XM_008760324.1 XP_008758546.1 1 1233

Gene Symbol
LOC103690131
Entrez Gene ID
103690131
Gene Name
tumor protein D55-like
Gene Aliases
Chromosome Location
Chr. 1: 248646221 - 248649734 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231260.2 XP_006231322.1 1233
XM_008760353.1 XP_008758575.1 1232

View Full Product Details