Gene Transcripts

Gene Symbol
Lhfpl5
Entrez Gene ID
294303
Gene Name
lipoma HMGIC fusion partner-like 5
Gene Aliases
Tmhs
Chromosome Location
Chr. 20: 5815837 - 5826137 on Build Rnor_6.0
UniGene ID
Rn.161732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013906.1 NP_001013928.1 1 475
GenBank mRNA BC087673.1 - 1 475

View Full Product Details