Gene Transcripts

Gene Symbol
NELFA
Entrez Gene ID
7469
Gene Name
negative elongation factor complex member A
Gene Aliases
NELF-A, P/OKcl.15, WHSC2
Chromosome Location
Chr. 4: 1982714 - 2009972 on Build GRCh38
UniGene ID
Hs.21771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005663.4 NP_005654.3 11 1720
XM_017008589.1 XP_016864078.1 12 2541
GenBank mRNA AB044549.1 - 10 1507
AF101434.1 - 11 1449
AF131751.1 - 7 970
AK001304.1 - 11 1507
AK094040.1 - 10 1280
AK095737.1 - 10 1763
AK126056.1 - 11 2595
AK222853.1 - 11 1457
BC002764.2 - 11 1460 3630594

View Full Product Details