Gene Transcripts

Gene Symbol
Mrps36
Entrez Gene ID
294696
Gene Name
mitochondrial ribosomal protein S36
Gene Aliases
Chromosome Location
Chr. 2: 30740618 - 30750901 on Build Rnor_6.0
UniGene ID
Rn.3406
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191605.3 NP_001178534.1 3 299
XM_006231846.2 XP_006231908.1 3 423
GenBank mRNA FM072467.1 - 3 299
FQ224224.1 - 3 233

View Full Product Details