Gene Transcripts

Gene Symbol
WNT8A
Entrez Gene ID
7478
Gene Name
Wnt family member 8A
Gene Aliases
WNT8D
Chromosome Location
Chr. 5: 138077367 - 138092900 on Build GRCh38
UniGene ID
Hs.591274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_058244.3 NP_490645.1 1, 2 270
NR_125351.1 1, 2 270
GenBank mRNA AB057725.1 - 1, 2 77
AY009402.1 - 1, 2 172

View Full Product Details