Gene Transcripts

Gene Symbol
Ube2q1
Entrez Gene ID
295252
Gene Name
ubiquitin-conjugating enzyme E2Q family member 1
Gene Aliases
Ube2q
Chromosome Location
Chr. 2: 189106039 - 189115110 on Build Rnor_6.0
UniGene ID
Rn.19034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106448.2 NP_001099918.1 1, 2 414
GenBank mRNA BC166534.1 - 1, 2 414

View Full Product Details