Gene Transcripts

Gene Symbol
Trim45
Entrez Gene ID
295323
Gene Name
tripartite motif-containing 45
Gene Aliases
Chromosome Location
Chr. 2: 203301838 - 203310532 on Build Rnor_6.0
UniGene ID
Rn.102590
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106453.1 NP_001099923.1 4, 5 1919

View Full Product Details