Gene Transcripts

Gene Symbol
Gstm6l
Entrez Gene ID
295362
Gene Name
glutathione S-transferase, mu 6-like
Gene Aliases
Gstm6
Chromosome Location
Chr. 2: 210698155 - 210703265 on Build Rnor_6.0
UniGene ID
Rn.136894
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106464.1 NP_001099934.1 6, 7 533

View Full Product Details