Gene Transcripts

Gene Symbol
Lppr4
Entrez Gene ID
295401
Gene Name
lipid phosphate phosphatase-related protein type 4
Gene Aliases
Prg-1, Prg1
Chromosome Location
Chr. 2: 220298239 - 220341863 on Build Rnor_6.0
UniGene ID
Rn.58355
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001508.2 NP_001001508.2 6 1491
GenBank mRNA AF541280.1 - 6 1492
AY266268.1 - 6 1492

View Full Product Details