Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282795.1 | NP_001269724.1 | 5 | 1793 | |
NM_001282797.1 | NP_001269726.1 | 4 | 1633 | ||
NM_003416.3 | NP_003407.1 | 5 | 1760 | ||
XM_006716654.2 | XP_006716717.1 | 3 | 1524 | ||
XM_006716656.3 | XP_006716719.1 | 3 | 1513 | ||
XM_011517291.2 | XP_011515593.1 | 5 | 1807 | ||
XM_011517292.2 | XP_011515594.1 | 5 | 2141 | ||
XM_011517293.2 | XP_011515595.1 | 5 | 2140 | ||
XM_011517294.2 | XP_011515596.1 | 5 | 1919 | ||
XM_011517295.2 | XP_011515597.1 | 5 | 1700 | ||
XM_011517296.2 | XP_011515598.1 | 5 | 1711 | ||
XM_011517297.2 | XP_011515599.1 | 3 | 1560 | ||
XM_017013813.1 | XP_016869302.1 | 5 | 1804 | ||
XM_017013814.1 | XP_016869303.1 | 4 | 2106 | ||
XM_017013815.1 | XP_016869304.1 | 4 | 2103 | ||
GenBank mRNA | AB209619.1 | - | 2 | 5149 | |
AK300001.1 | - | 5 | 1728 | ||
AK301561.1 | - | 4 | 1594 | ||
AK313745.1 | - | 5 | 1700 | ||
BC058923.1 | - | 5 | 1704 | 4516166 | |
BX647998.1 | - | 5 | 1719 | ||
M29580.1 | - | 5 | 1861 |