Gene Transcripts

Gene Symbol
Rsb66
Entrez Gene ID
296610
Gene Name
Rsb-66 protein
Gene Aliases
Chromosome Location
Chr. 3: 7269851 - 7279356 on Build Rnor_6.0
UniGene ID
Rn.46431
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181694.2 NP_859045.2 3, 4 423
XM_006233762.1 XP_006233824.1 3, 4 586
XM_006233763.2 XP_006233825.1 3, 4 802
XM_006233764.2 XP_006233826.1 3, 4 1257
XM_006233766.1 XP_006233828.1 3, 4 586
GenBank mRNA AY121839.1 - 2, 3 373
BC097268.1 - 3, 4 423 7457912

View Full Product Details