Gene Transcripts

Gene Symbol
Podxl2
Entrez Gene ID
297433
Gene Name
podocalyxin-like 2
Gene Aliases
RGD1305815
Chromosome Location
Chr. 4: 120785494 - 120817335 on Build Rnor_6.0
UniGene ID
Rn.100042
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106607.1 NP_001100077.1 3 1125
GenBank mRNA BC161970.1 - 3 1167

View Full Product Details