Gene Transcripts
- Gene Symbol
- V1ra14
- Entrez Gene ID
- 297442
- Gene Name
- vomeronasal 1 receptor A14
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.215804
- Species
- Rattus norvegicus
- Gene Symbol
- Vom1r103
- Entrez Gene ID
- 100312735
- Gene Name
- vomeronasal 1 receptor 103
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.144688
- Species
- Rattus norvegicus
- Gene Symbol
- Vom1r105
- Entrez Gene ID
- 286893
- Gene Name
- vomeronasal 1 receptor 105
- Gene Aliases
- Vmn1r51, Vn1, Vnr1
- Chromosome Location
- Chr. 4: 122124451 - 122131893 on Build Rnor_6.0
- UniGene ID
- Rn.144688
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173113.1 |
NP_775136.1
|
2 |
1075 |
|
GenBank mRNA |
U36785.1 |
- |
2 |
1075 |
|
View Full Product Details