Gene Transcripts

Gene Symbol
Casc1
Entrez Gene ID
297720
Gene Name
cancer susceptibility candidate 1
Gene Aliases
Las1
Chromosome Location
Chr. 4: 179440984 - 179476932 on Build Rnor_6.0
UniGene ID
Rn.162764
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106627.3 NP_001100097.2 14 1765
XM_008763378.1 XP_008761600.1 10 2202
GenBank mRNA BC160907.1 - 14 1765
DV722478.1 - 2 252

View Full Product Details