Gene Transcripts

Gene Symbol
Arid1a
Entrez Gene ID
297867
Gene Name
AT rich interactive domain 1A (SWI-like)
Gene Aliases
Chromosome Location
Chr. 5: 151904687 - 151977966 on Build Rnor_6.0
UniGene ID
Rn.61077
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106635.1 NP_001100105.1 19 4092
XM_006239011.2 XP_006239073.1 20 5289
XM_006239012.2 XP_006239074.1 20 5286

View Full Product Details