Gene Transcripts

Gene Symbol
Zbtb8os
Entrez Gene ID
297885
Gene Name
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases
RGD1562249, Zbtb8os-ps1
Chromosome Location
Chr. 5: 147535585 - 147547252 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750036.3 XP_003750084.3 4, 5 766

View Full Product Details