Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318144.1 | NP_001305073.1 | 3 | 404 | |
NM_004925.4 | NP_004916.1 | 3 | 404 | ||
GenBank mRNA | AB001325.1 | - | 3 | 361 | |
AJ493597.1 | - | 3 | 324 | ||
AK095363.1 | - | 3 | 674 | ||
AK292208.1 | - | 4 | 364 | ||
AK303206.1 | - | 3 | 364 | ||
AK315760.1 | - | 3 | 364 | ||
BC013566.1 | - | 3 | 338 | 3877096 | |
BT007199.1 | - | 3 | 301 | ||
CR541991.1 | - | 4 | 301 | ||
CR542025.1 | - | 4 | 301 | ||
DA976040.1 | - | 3 | 404 |