Gene Transcripts

Gene Symbol
Tbk1
Entrez Gene ID
299827
Gene Name
TANK-binding kinase 1
Gene Aliases
Chromosome Location
Chr. 7: 63655247 - 63687978 on Build Rnor_6.0
UniGene ID
Rn.30683
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106786.1 NP_001100256.1 7 896

View Full Product Details