Gene Transcripts

Gene Symbol
AQP4
Entrez Gene ID
361
Gene Name
aquaporin 4
Gene Aliases
MIWC, WCH4
Chromosome Location
Chr. 18: 26852038 - 26865844 on Build GRCh38
UniGene ID
Hs.315369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317384.1 NP_001304313.1 4, 5 844
NM_001650.5 NP_001641.1 4, 5 844
NM_004028.4 NP_004019.1 3, 4 971
XM_011525942.2 XP_011524244.1 4, 5 886
GenBank mRNA AB128929.1 - 5, 6 374
AK222684.1 - 4, 5 756
AK295069.1 - 5, 6 655
BC022286.1 - 4, 5 709 4717755
D63412.1 - 4, 5 709
HQ901095.1 - 4, 5 818
U34845.1 - 3, 4 907
U63622.1 - 4, 5 728
U63623.1 - 3, 4 632

View Full Product Details