Gene Transcripts

Gene Symbol
Phf21b
Entrez Gene ID
300117
Gene Name
PHD finger protein 21B
Gene Aliases
RGD1308739
Chromosome Location
Chr. 7: 125649688 - 125720694 on Build Rnor_6.0
UniGene ID
Rn.198794
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130680.1 NP_001124152.1 6 1149
XM_008765685.1 XP_008763907.1 6 736

View Full Product Details