Gene Transcripts

Gene Symbol
Lman1l
Entrez Gene ID
300743
Gene Name
lectin, mannose-binding, 1 like
Gene Aliases
slamp
Chromosome Location
Chr. 8: 62386806 - 62399648 on Build Rnor_6.0
UniGene ID
Rn.163005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012465.1 NP_001012483.1 1 87
GenBank mRNA AB188302.1 - 1 87

View Full Product Details