Gene Transcripts

Gene Symbol
Mettl21c
Entrez Gene ID
301378
Gene Name
methyltransferase like 21C
Gene Aliases
RGD1561425
Chromosome Location
Chr. 9: 50750971 - 50762082 on Build Rnor_6.0
UniGene ID
Rn.37634
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106908.1 NP_001100378.1 5 509
XM_008767022.1 XP_008765244.1 4 1301
GenBank mRNA FQ215023.1 - 883

View Full Product Details