Gene Transcripts

Gene Symbol
REEP5
Entrez Gene ID
7905
Gene Name
receptor accessory protein 5
Gene Aliases
C5orf18, D5S346, DP1, TB2, YOP1, Yip2e
Chromosome Location
Chr. 5: 112876384 - 112922334 on Build GRCh38
UniGene ID
Hs.429608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005669.4 NP_005660.4 3 377
XM_017009844.1 XP_016865333.1 3 294
GenBank mRNA AK294496.1 - 2 363
AK298268.1 - 3 281
AK299902.1 - 3 273
AK310729.1 - 2 373
AK312522.1 - 3 282
AY562243.1 - 3 233
BC000232.1 - 3 258 3350749
BC065926.1 - 3 236 6164621
BX648005.1 - 3 358
DA807043.1 - 3 378
M74090.1 - 3 259

View Full Product Details