Gene Transcripts

Gene Symbol
Tmem8a
Entrez Gene ID
303004
Gene Name
transmembrane protein 8A
Gene Aliases
M83, Tmem8
Chromosome Location
Chr. 10: 15484951 - 15495380 on Build Rnor_6.0
UniGene ID
Rn.21890
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106991.1 NP_001100461.1 6 1302
XM_006245973.2 XP_006246035.1 6 1916
XM_008767577.1 XP_008765799.1 6 2994

View Full Product Details