Gene Transcripts

Gene Symbol
Wnt3a
Entrez Gene ID
303181
Gene Name
wingless-type MMTV integration site family, member 3A
Gene Aliases
Chromosome Location
Chr. 10: 45598898 - 45643151 on Build Rnor_6.0
UniGene ID
Rn.234269
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107005.2 NP_001100475.2 4 748
XM_006246487.2 XP_006246549.1 4 906

View Full Product Details