Gene Transcripts

Gene Symbol
Med13
Entrez Gene ID
303403
Gene Name
mediator complex subunit 13
Gene Aliases
Thrap1
Chromosome Location
Chr. 10: 73698789 - 73787084 on Build Rnor_6.0
UniGene ID
Rn.94023
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107035.1 NP_001100505.1 2 324
XM_006247066.2 XP_006247128.1 2 328

View Full Product Details