Gene Transcripts

Gene Symbol
Tns4
Entrez Gene ID
303517
Gene Name
tensin 4
Gene Aliases
RGD1310402
Chromosome Location
Chr. 10: 86977504 - 87001256 on Build Rnor_6.0
UniGene ID
Rn.23329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024881.1 NP_001020052.1 2 525
GenBank mRNA BC097378.1 - 2 525

View Full Product Details