Gene Transcripts

Gene Symbol
PSCA
Entrez Gene ID
8000
Gene Name
prostate stem cell antigen
Gene Aliases
PRO232
Chromosome Location
Chr. 8: 142670308 - 142682727 on Build GRCh38
UniGene ID
Hs.652235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005672.4 NP_005663.2 3 268
NR_033343.1 3 421
GenBank mRNA AF043498.1 - 3 229
AJ297436.1 - 3 231
AY358912.1 - 3 201
BC023582.2 - 3 218 4840974
BC048808.1 - 3 218
BC065183.1 - 3 218 5557096
BM768967.1 - 3 269
BU157227.1 - 3 408
CB993163.1 - 3 295

View Full Product Details