Gene Transcripts

Gene Symbol
Trim65
Entrez Gene ID
303684
Gene Name
tripartite motif-containing 65
Gene Aliases
RGD1559497
Chromosome Location
Chr. 10: 104675950 - 104681484 on Build Rnor_6.0
UniGene ID
Rn.29932
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135714.1 NP_001129186.1 5, 6 1510
GenBank mRNA FQ223610.1 - 1081

View Full Product Details