Gene Transcripts

Gene Symbol
Fscn2
Entrez Gene ID
303741
Gene Name
fascin actin-bundling protein 2, retinal
Gene Aliases
Chromosome Location
Chr. 10: 109528330 - 109540026 on Build Rnor_6.0
UniGene ID
Rn.138713
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107072.1 NP_001100542.1 3 1169
XM_008768476.1 XP_008766698.1 3 6326

View Full Product Details